accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOOH3C

CAS RN 13575-74-1

IUPAC Name: 4-(3,4-dimethoxyphenyl)butanoic acid
Synonyms: 4-(3,4-dimethoxyphenyl)butanoic acid
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: IDQKCIAJGRMMNC-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCCC(O)=O)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results