accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135788-08-8

IUPAC Name: 2,3-bis(4-chlorophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 2,3-bis(4-chlorophenyl)-5-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 403.69
Molecular Formula: C19H13Cl3N4
InChi Key: AUTAPTQTFVZDGP-UHFFFAOYSA-M
SMILES: [Cl-].ClC1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=C(Cl)C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results