Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(4-chlorophenyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. B1052100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1052100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1052100MG Supplier TCI America Supplier No. B1052100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135788-08-8
Molecular Formula C19H13Cl3N4
Molecular Weight (g/mol) 403.691
MDL Number MFCD00060008
InChI Key AUTAPTQTFVZDGP-UHFFFAOYSA-M
PubChem CID 44630120
IUPAC Name 2,3-bis(4-chlorophenyl)-5-phenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=C(C=C3)Cl)C4=CC=C(C=C4)Cl.[Cl-]

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 403.69
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-chlorophenyl)-5-phenyltetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.