accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13587-26-3

NH3NH3H2OH2OH2OH2OH2OH2OOHSOOOOHSOOOCu

CAS RN 13587-26-3

IUPAC Name: copper(2+) diamine hexahydrate dihydrogen sulfate
Synonyms: copper(2+) diamine hexahydrate dihydrogen sulfate
Molecular Weight (g/mol): 399.83
Molecular Formula: CuH20N2O14S2
InChi Key: IHEZHABINDDXAE-UHFFFAOYSA-L
SMILES: N.N.O.O.O.O.O.O.[Cu++].OS([O-])(=O)=O.OS([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results