accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3ONN

CAS RN 13593-03-8

IUPAC Name: O,O-diethyl O-quinoxalin-2-yl phosphorothioate
Synonyms: savall sandoz 6538 spencer S-6538 chinalphos diethquinalphione diethquinalphion wie oben ekalux 25EC diethylquinalphione quinalphos 25 EC
Molecular Weight (g/mol): 298.3
Molecular Formula: C12H15N2O3PS
InChi Key: JYQUHIFYBATCCY-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CN=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results