accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 136-45-8

IUPAC Name: 2,5-dipropyl pyridine-2,5-dicarboxylate
Synonyms: dipropyl pyridinedicarboxylate
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: IITCWRFYJWUUPC-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=C(N=C1)C(=O)OCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results