accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13600-43-6

IUPAC Name: 4-(trifluoromethyl)pyridine-3-carbonitrile
Synonyms: 4-(trifluoromethyl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 172.11
Molecular Formula: C7H3F3N2
InChi Key: DHIRCRHQLUNYDS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=NC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results