Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)nicotinonitrile, ≥95%, Thermo Scientific™

Catalog No. RF04123EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RF04123EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF04123EA Supplier Thermo Scientific Chemicals Supplier No. RF04123EA
Add to Cart

Chemical Identifiers

CAS 13600-43-6
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD00153107
InChI Key DHIRCRHQLUNYDS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl nicotinonitrile, 3-cyano-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridine-3-carbonitrile, 3-cyano-4-trifluoromethylpyridine, 4-trifluoromethyl-3-pyridinenitrile, 3-pyridinecarbonitrile, 4-trifluoromethyl, 4-trifluoromethyl-nicotinonitrile, pubchem9232, buttpark 4357-45, ksc495c9t
PubChem CID 2736743
IUPAC Name 4-(trifluoromethyl)pyridine-3-carbonitrile
SMILES FC(F)(F)C1=C(C=NC=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 172.11
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)nicotinonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.