accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CN

CAS RN 13602-11-4

IUPAC Name: methyl 6-methylpyridine-2-carboxylate
Synonyms: methyl 6-methylpyridine-2-carboxylate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: CYWIZMOZBTXFIL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results