Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Methylpyridine-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M25035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25035G Supplier TCI America Supplier No. M25035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13602-11-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD06203670
InChI Key CYWIZMOZBTXFIL-UHFFFAOYSA-N
Synonym methyl 6-methylpicolinate, 6-methylpyridine-2-carboxylic acid methyl ester, methyl 6-methyl-2-pyridinecarboxylate, 6-methyl-pyridine-2-carboxylic acid methyl ester, 6-methylpicolinic acid methyl ester, 2-pyridinecarboxylic acid, 6-methyl-, methyl ester, pubchem20955, methyl-6-methylpicolinate, ksc495e0b, aronis24547
PubChem CID 73022
IUPAC Name methyl 6-methylpyridine-2-carboxylate
SMILES COC(=O)C1=CC=CC(C)=N1

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 6-Methylpyridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.