accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OO

CAS RN 13612-34-5

IUPAC Name: 2-(2,5-dimethylphenyl)acetate
Synonyms: (2,5-dimethylphenyl)acetate
Molecular Weight (g/mol): 163.2
Molecular Formula: C10H11O2
InChi Key: RUSCTNYOPQOXDJ-UHFFFAOYSA-M
SMILES: CC1=CC=C(C)C(CC([O-])=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)