Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D377925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D377925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D377925G Supplier TCI America Supplier No. D377925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical reagent with >98.0% for laboratory research purposes.

Chemical Identifiers

CAS 13612-34-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00014369
InChI Key RUSCTNYOPQOXDJ-UHFFFAOYSA-N
Synonym 2,5-dimethylphenylacetic acid, 2-2,5-dimethylphenyl acetic acid, 2,5-dimethylphenyl acetic acid, 2,5-xylylacetic acid, 2,5-dimethylphenylaceticacid, 2,5-dimethylbenzeneacetic acid, benzeneacetic acid, 2,5-dimethyl, pubchem22525, acmc-1bb4h
PubChem CID 83615
IUPAC Name 2-(2,5-dimethylphenyl)acetic acid
SMILES CC1=CC(=C(C=C1)C)CC(=O)O

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Chemical Name or Material 2,5-Dimethylphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.