CAS RN 136207-26-6
CAS RN 136207-26-6
IUPAC Name:
(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Synonyms:
(2R)-2-[(tert-butoxycarbonyl)amino]-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Molecular Weight (g/mol):
355.26
Molecular Formula:
C14H14F5NO4
InChi Key:
UZDKQMIDSLETST-ZCFIWIBFSA-N
SMILES:
CC(C)(C)OC(=O)N[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Phe F5 -OH, Advanced ChemTech
CAS: 136207-26-6 Molecular Formula: C14H14F5NO4 Molecular Weight (g/mol): 355.26 MDL Number: MFCD00151892 InChI Key: UZDKQMIDSLETST-ZCFIWIBFSA-N IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid SMILES: CC(C)(C)OC(=O)N[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)C(O)=O