CAS RN 136259-20-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals P32/98, 98%
Thermo Scientific Chemicals P32/98, 98%
CAS: 136259-20-6 Molecular Formula: C9H18N2OS.0.5C4H4O4 Synonym: 3-[(2S,3S)-2-Amino-3-methylpentanoyl]thiazolidine hemifumarate;