accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3NNH2NNH3NOHNO

CAS RN 136587-13-8

IUPAC Name: N-[(3-azaniumylpropyl)({4-[(3-azaniumylpropyl)azaniumyl]butyl})amino]-N-nitrosohydroxylamine
Synonyms: N-[(3-ammoniopropyl)({4-[(3-ammoniopropyl)ammonio]butyl})amino]-N-nitrosohydroxylamine
Molecular Weight (g/mol): 265.38
Molecular Formula: C10H29N6O2
InChi Key: QPIOUFBAROSIAN-UHFFFAOYSA-Q
SMILES: [NH3+]CCC[NH2+]CCCCN(CCC[NH3+])N(O)N=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results