Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermine NONOate 97%, Thermo Scientific™

Catalog No. AC328860250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328860250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328860250 Supplier Thermo Scientific Chemicals Supplier No. 328860250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136587-13-8
Molecular Formula C10H26N6O2
Molecular Weight (g/mol) 262.358
InChI Key QPIOUFBAROSIAN-UHFFFAOYSA-N
Synonym spermine nonoate, spermine-nitric oxide, spermine nitric oxide complex, noc 22, noc-22, spermine nitric oxide adduct, spermine/nitric oxide complex, ccris 4091, sper/no, sper-no
PubChem CID 5259
IUPAC Name N-[3-aminopropyl-[4-(3-aminopropylamino)butyl]amino]-N-hydroxynitrous amide
SMILES C(CCN(CCCN)N(N=O)O)CNCCCN

Specifications

Melting Point 85°C to 95°C
Color White
Infrared Spectrum Authentic
Quantity 25 mg
Formula Weight 262.4
Percent Purity ≥96.0%
Physical Form Crystalline Solid
Chemical Name or Material Spermine NONOate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.