accessibility menu, dialog, popup

UserName

Viewing profile as user:
SFFFFOOOOOOOOOHNNHNHNNN(R)(R)CH3(S)(R)(S)CH3CH3H3C(R)(R)HHHH(Z)

CAS RN 1365970-03-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results