Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glecaprevir, Thermo Scientific™

Catalog No. AC469341000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469341000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469341000 Supplier Thermo Scientific Chemicals Supplier No. 469341000
Add to Cart
This item is not returnable. View return policy

Specifications

Color White to Almost White
Description Glecaprevir
CAS 1365970-03-1
Molecular Formula C38 H46 F4 N6 O9 S
Shipping Condition No special requirements
Product Line Thermo Scientific
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.