accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PPH3CH3CH3C

CAS RN 136705-62-9

IUPAC Name: 1-[2-(2,5-diethylphospholan-1-yl)ethyl]-2,5-diethylphospholane
Synonyms: 1-[2-(2,5-diethylphospholan-1-yl)ethyl]-2,5-diethylphospholane
Molecular Weight (g/mol): 314.43
Molecular Formula: C18H36P2
InChi Key: QOLRLVPABLMMKI-UHFFFAOYNA-N
SMILES: CCC1CCC(CC)P1CCP1C(CC)CCC1CC
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results