accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSO

CAS RN 13673-63-7

IUPAC Name: 3-methyl-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 3-methyl-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: VMOIPIVQHVFUFO-UHFFFAOYSA-N
SMILES: CN1C(=S)OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results