accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSH3C

CAS RN 13679-72-6

IUPAC Name: 1-(3-methylthiophen-2-yl)ethan-1-one
Synonyms: 1-(3-methylthiophen-2-yl)ethanone
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: YBJDKNXEWQSGEL-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C=CS1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results