accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)NONH(R)N(R)NO(R)OOOCH3OH3CPONCH3CH3H3CCH3NF

CAS RN 136834-22-5

IUPAC Name: N-{9-[(2R,3R,4R,5R)-4-[bis(4-methoxyphenyl)(phenyl)methoxy]-5-[({[bis(propan-2-yl)amino](2-cyanoethoxy)phosphanyl}oxy)methyl]-3-fluorooxolan-2-yl]-9H-purin-6-yl}benzamide
Synonyms: N-{9-[(2R,3R,4R,5R)-4-[bis(4-methoxyphenyl)(phenyl)methoxy]-5-({[(2-cyanoethoxy)(diisopropylamino)phosphanyl]oxy}methyl)-3-fluorooxolan-2-yl]purin-6-yl}benzamide
Molecular Weight (g/mol): 875.94
Molecular Formula: C47H51FN7O7P
InChi Key: NZDMBUCNULHYQM-JRJFZAHINA-N
SMILES: COC1=CC=C(C=C1)C(O[C@@H]1[C@@H](COP(OCCC#N)N(C(C)C)C(C)C)O[C@H]([C@@H]1F)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results