Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite), 98%

Catalog No. AAJ6406403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6406403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6406403 Supplier Thermo Scientific Chemicals Supplier No. J6406403
  • $191.20
    $162.65 / Each of 1

    Save $28.55

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 136834-22-5 | C47H51FN7O7P | 875.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136834-22-5
Molecular Formula C47H51FN7O7P
Molecular Weight (g/mol) 875.94
MDL Number MFCD12912392
InChI Key NZDMBUCNULHYQM-JRJFZAHINA-N
Synonym 2r,3r,4r,5r-5-6-benzamido-9h-purin-9-yl-2-bis 4-methoxyphenyl phenyl methoxy methyl-4-fluorotetrahydrofuran-3-yl 2-cyanoethyl diisopropylphosphoramidite, dmt-2'fluoro-da bz phosphoramidite, dmt-2'fluoro-da bz phosphoramidite, configured for abi, dmt-2'fluoro-da bz phosphoramidite, configured for perkinelmer, configured for polygen, 2'-f-bz-ra, dmt-2'fluoro-da bz amidite, 2'-fluoro-n6-bz-5'-o-dmt-2'-da-3'-ce-phosphoramidite, dmt-2'fluoro-da bz phosphoramidite, configured for ??kta r and oligopilot r, n-9-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-2-cyanoethoxy diisopropylamino phosphanyl oxy-3-fluorooxolan-2-yl purin-6-yl benzamide, n-benzoyl-5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-2'-fluoroadenosine 3'-2-cyanoethyl n,n-bis 1-methylethyl phosphoramidite
PubChem CID 12042896
IUPAC Name N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-[2-cyanoethoxy-[di(propan-2-yl)amino]phosphanyl]oxy-3-fluorooxolan-2-yl]purin-6-yl]benzamide
SMILES COC1=CC=C(C=C1)C(O[C@@H]1[C@@H](COP(OCCC#N)N(C(C)C)C(C)C)O[C@H]([C@@H]1F)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Color White
Quantity 1 g
Formula Weight 875.92
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosp
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.