accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFPFFFCl

CAS RN 13685-24-0

IUPAC Name: chlorobis[4-(trifluoromethyl)phenyl]phosphane
Synonyms: chlorobis[4-(trifluoromethyl)phenyl]phosphane
Molecular Weight (g/mol): 356.63
Molecular Formula: C14H8ClF6P
InChi Key: PBLUOOURLNAOAJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)P(Cl)C1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results