Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%

Catalog No. AAH2588806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2588806 5 g
AAH2588803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2588806 Supplier Thermo Scientific Chemicals Supplier No. H2588806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13685-24-0 | C14H8ClF6P | 356.632 g/mol

Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13685-24-0
Molecular Formula C14H8ClF6P
Molecular Weight (g/mol) 356.632
MDL Number MFCD04972304
InChI Key PBLUOOURLNAOAJ-UHFFFAOYSA-N
Synonym bis 4-trifluoromethylphenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphine, bis 4-trifluoromethyl phenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphane, acmc-20alnb, chloro-bis 4-trifluoromethyl phenyl phosphane
PubChem CID 4379933
IUPAC Name chloro-bis[4-(trifluoromethyl)phenyl]phosphane
SMILES C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)Cl

Specifications

Boiling Point 110°C (0.4 mmHg)
Quantity 5 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 356.64
Percent Purity 97%
Chemical Name or Material Chlorobis[4-(trifluoromethyl)phenyl]phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.