CAS RN 136861-46-6
IUPAC Name:
1-methyl-4-{[(4-{[(4-methylphenyl)methoxy]methyl}phenyl)methoxy]methyl}benzene
Synonyms:
1-methyl-4-{[(4-{[(4-methylphenyl)methoxy]methyl}phenyl)methoxy]methyl}benzene
Molecular Weight (g/mol):
346.47
Molecular Formula:
C24H26O2
InChi Key:
IJMQICMKIDWWAK-UHFFFAOYSA-N
SMILES:
CC1=CC=C(COCC2=CC=C(COCC3=CC=C(C)C=C3)C=C2)C=C1
1
–
1
of
1
results
1
alpha,alpha'-Bis(4-methylbenzyloxy)-p-xylene 98.0+%, TCI America™
CAS: 136861-46-6 Molecular Formula: C24H26O2 Molecular Weight (g/mol): 346.47 MDL Number: MFCD00143577 InChI Key: IJMQICMKIDWWAK-UHFFFAOYSA-N Synonym: p-Xylylene Glycol Bis(4-methylbenzyl) Ether PubChem CID: 21659525 IUPAC Name: 1,4-bis[(4-methylphenyl)methoxymethyl]benzene SMILES: CC1=CC=C(C=C1)COCC2=CC=C(C=C2)COCC3=CC=C(C=C3)C