Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Bis(4-methylbenzyloxy)-p-xylene 98.0+%, TCI America™
SDP

Catalog No. B14825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14825G 5 g
B14821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14825G Supplier TCI America Supplier No. B14825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 136861-46-6
Molecular Formula C24H26O2
Molecular Weight (g/mol) 346.47
MDL Number MFCD00143577
InChI Key IJMQICMKIDWWAK-UHFFFAOYSA-N
Synonym p-Xylylene Glycol Bis(4-methylbenzyl) Ether
PubChem CID 21659525
IUPAC Name 1,4-bis[(4-methylphenyl)methoxymethyl]benzene
SMILES CC1=CC=C(C=C1)COCC2=CC=C(C=C2)COCC3=CC=C(C=C3)C

Specifications

Melting Point 100°C
Color White
Quantity 5 g
Formula Weight 346.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Bis(4-methylbenzyloxy)-p-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.