CAS RN 136888-21-6
IUPAC Name:
2-chloro-5-fluoro-3-nitropyridine
Synonyms:
2-chloro-5-fluoro-3-nitropyridine
Molecular Weight (g/mol):
176.53
Molecular Formula:
C5H2ClFN2O2
InChi Key:
SVVZGNAZMPSGMU-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(F)=CN=C1Cl
1
–
1
of
1
results
1
2-Chloro-5-fluoro-3-nitropyridine 98.0+%, TCI America™
CAS: 136888-21-6 Molecular Formula: C5H2ClFN2O2 Molecular Weight (g/mol): 176.531 MDL Number: MFCD06659490 InChI Key: SVVZGNAZMPSGMU-UHFFFAOYSA-N PubChem CID: 10773626 IUPAC Name: 2-chloro-5-fluoro-3-nitropyridine SMILES: C1=C(C(=NC=C1F)Cl)[N+](=O)[O-]