Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluoro-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. C3115200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3115200MG 200 mg
C31151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3115200MG Supplier TCI America Supplier No. C3115200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 136888-21-6
Molecular Formula C5H2ClFN2O2
Molecular Weight (g/mol) 176.531
MDL Number MFCD06659490
InChI Key SVVZGNAZMPSGMU-UHFFFAOYSA-N
PubChem CID 10773626
IUPAC Name 2-chloro-5-fluoro-3-nitropyridine
SMILES C1=C(C(=NC=C1F)Cl)[N+](=O)[O-]

Specifications

Melting Point 50°C
Color Yellow
Quantity 200 mg
Synonym 2-chloro-3-nitro-5-fluoropyridine, 2-chloro-5-fluoro-3-nitro-pyridine, pyridine, 2-chloro-5-fluoro-3-nitro, pubchem2967, acmc-1cc17, 2-chloro-5-fluoro-3-nitro pyridine, pyridine,2-chloro-5-fluoro-3-nitro
Formula Weight 176.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-fluoro-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.