accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2COOFFFFFFF

CAS RN 13695-31-3

IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutyl 2-methylprop-2-enoate
Synonyms: 2,2,3,3,4,4,4-heptafluorobutyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 268.13
Molecular Formula: C8H7F7O2
InChi Key: VIEHKBXCWMMOOU-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results