Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.

Catalog No. AAL1292214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1292214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1292214 Supplier Thermo Scientific Chemicals Supplier No. L1292214
  • $457.00
    $388.65 / Each of 1

    Save $68.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13695-31-3 | C8H7F7O2 | 268.131 g/mol

2,2,3,3,4,4,4-Heptafluorobutyl methacrylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13695-31-3
Molecular Formula C8H7F7O2
Molecular Weight (g/mol) 268.131
MDL Number MFCD00039251
InChI Key VIEHKBXCWMMOOU-UHFFFAOYSA-N
Synonym heptafluorobutyl methacrylate, 2,2,3,3,4,4,4-heptafluorobutyl methacrylate, 1h,1h-heptafluorobutyl methacrylate, 1h,1h-heptafluoro-n-butyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,4-heptafluorobutyl ester, methacrylic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, methacrylic acid 2,2,3,3,4,4,4-heptafluorobutyl ester, heptafluoropropylmethyl methacrylate, 2,2,3,3,4,4,4-heptafluorobutyl 2-methylacrylate #
PubChem CID 83666
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.345
Boiling Point 134°C to 136°C
Flash Point 36°C (96°F)
Odor Recognizable
Refractive Index 1.342
Quantity 25 g
UN Number UN3272
Solubility Information Not miscible in water.
Formula Weight 268.13
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, stabilized
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 237-217-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.