accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13697-89-7

IUPAC Name: 1,3-difluoro-2-iodobenzene
Synonyms: 1,3-difluoro-2-iodobenzene
Molecular Weight (g/mol): 239.99
Molecular Formula: C6H3F2I
InChi Key: VQMXWPLTZBKNEH-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results