Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Difluoro-2-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. D36435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36435G Supplier TCI America Supplier No. D36435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13697-89-7
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.991
MDL Number MFCD04039247
InChI Key VQMXWPLTZBKNEH-UHFFFAOYSA-N
PubChem CID 2782738
IUPAC Name 1,3-difluoro-2-iodobenzene
SMILES C1=CC(=C(C(=C1)F)I)F

Specifications

Color White-Yellow
Boiling Point 71°C
Quantity 5 g
Formula Weight 239.99
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,3-Difluoro-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.