accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHClCl

CAS RN 137-19-9

IUPAC Name: 4,6-dichlorobenzene-1,3-diol
Synonyms: 1,3-benzenediol, 4,6-dichloro-
Molecular Weight (g/mol): 179
Molecular Formula: C6H4Cl2O2
InChi Key: GRLQBYQELUWBIO-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C(Cl)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results