Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dichlororesorcinol 96.0+%, TCI America™
SDP

Catalog No. D041325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D041325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D041325G Supplier TCI America Supplier No. D041325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 137-19-9
Molecular Formula C6H4Cl2O2
Molecular Weight (g/mol) 178.996
MDL Number MFCD00002274
InChI Key GRLQBYQELUWBIO-UHFFFAOYSA-N
Synonym 4,6-dichlororesorcinol, 1,3-benzenediol, 4,6-dichloro, 4,6-dichloro-1,3-benzenediol, 4,6-dichloro-1,3-dihydroxybenzene, resorcinol, 4,6-dichloro, 1,5-dichloro-2,4-dihydroxy-benzene, 4,3-dihydroxybenzene, 3,4-dichlororesorcinol, resorcinol,6-dichloro, acmc-209caz
PubChem CID 67299
IUPAC Name 4,6-dichlorobenzene-1,3-diol
SMILES C1=C(C(=CC(=C1O)Cl)Cl)O

Specifications

Melting Point 115°C
Boiling Point 125°C
Quantity 25 g
UN Number 2811
Formula Weight 179.00
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichlororesorcinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.