CAS RN 137-66-6
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
CAS: 137-66-6 Molecular Formula: C22H38O7 Molecular Weight (g/mol): 414.54 MDL Number: MFCD00005377 InChI Key: NULLRXHRYOXSEW-UHFFFAOYNA-N Synonym: ascorbyl palmitate,l-ascorbyl 6-palmitate,6-o-palmitoyl-l-ascorbic acid,ascorbic palmitate,l-ascorbyl palmitate,unii-qn83us2b0n,ascorbyl monopalmitate,l-ascorbic acid 6-palmitate,s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate,ascorbyl palmitate nf PubChem CID: 54680660 IUPAC Name: [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
Ascorbyl Palmitate, NF, 95-100.5%, Spectrum™ Chemical
CAS: 137-66-6 Molecular Formula: C22H38O7 Molecular Weight (g/mol): 414.54 MDL Number: MFCD00005377 InChI Key: NULLRXHRYOXSEW-UHFFFAOYNA-N IUPAC Name: 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
Ascorbyl Palmitate, FCC, 95%, Spectrum™ Chemical
CAS: 137-66-6 Molecular Formula: C22H38O7 Molecular Weight (g/mol): 414.54 MDL Number: MFCD00005377 InChI Key: NULLRXHRYOXSEW-UHFFFAOYNA-N IUPAC Name: 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
L-Ascorbyl palmitate, MP Biomedicals™
CAS: 137-66-6 Molecular Formula: C22H38O7 Molecular Weight (g/mol): 414.54 MDL Number: MFCD00005377 InChI Key: NULLRXHRYOXSEW-UHFFFAOYNA-N Synonym: ascorbyl palmitate,l-ascorbyl 6-palmitate,6-o-palmitoyl-l-ascorbic acid,ascorbic palmitate,l-ascorbyl palmitate,unii-qn83us2b0n,ascorbyl monopalmitate,l-ascorbic acid 6-palmitate,s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate,ascorbyl palmitate nf PubChem CID: 54680660 IUPAC Name: 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
L-Ascorbic acid 6-palmitate, 99%
CAS: 137-66-6 Molecular Formula: C22H38O7 Molecular Weight (g/mol): 414.54 MDL Number: MFCD00005377 InChI Key: NULLRXHRYOXSEW-UHFFFAOYNA-N Synonym: ascorbyl palmitate,l-ascorbyl 6-palmitate,6-o-palmitoyl-l-ascorbic acid,ascorbic palmitate,l-ascorbyl palmitate,unii-qn83us2b0n,ascorbyl monopalmitate,l-ascorbic acid 6-palmitate,s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate,ascorbyl palmitate nf PubChem CID: 54680660 IUPAC Name: [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
6-O-Palmitoyl-L-ascorbic Acid 97.0+%, TCI America™
CAS: 137-66-6 Molecular Formula: C22H38O7 Molecular Weight (g/mol): 414.54 MDL Number: MFCD00005377 InChI Key: NULLRXHRYOXSEW-UHFFFAOYNA-N Synonym: ascorbyl palmitate,l-ascorbyl 6-palmitate,6-o-palmitoyl-l-ascorbic acid,ascorbic palmitate,l-ascorbyl palmitate,unii-qn83us2b0n,ascorbyl monopalmitate,l-ascorbic acid 6-palmitate,s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate,ascorbyl palmitate nf PubChem CID: 54680660 IUPAC Name: 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
Medisca Ascorbyl Palmitate NF
Ascorbyl Palmitate NF is a white to yellowish white powder, having a characteristic odor. Alternative name(s): Palmitoyl l-ascorbic