accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3CH3

CAS RN 137-89-3

IUPAC Name: 1,3-bis(2-ethylhexyl) benzene-1,3-dicarboxylate
Synonyms: bis(2-ethylhexyl) isophthalate
Molecular Weight (g/mol): 390.56
Molecular Formula: C24H38O4
InChi Key: WXZOXVVKILCOPG-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC(=CC=C1)C(=O)OCC(CC)CCCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results