Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) isophthalate, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018991 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8991 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8991 Supplier MilliporeSigma Supelco Supplier No. 91344100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 137-89-3
Physical Form Neat
Linear Formula C24H38O4
Synonym 1,3-Benzenedicarboxylic acid 1,3-bis(2-ethylhexyl) ester; DOIP; Di-2-ethylhexyl isophthalate; Isophthalic acid bis(2-ethylhexyl ester)
Type Analytical Standard
Quantity 100 mg
Formula Weight 390.56
Percent Purity ≥98% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.