accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3CH3

CAS RN 13702-58-4

IUPAC Name: 3,4,6-trimethylcyclohex-3-ene-1-carbaldehyde
Synonyms: 3,4,6-trimethylcyclohex-3-ene-1-carbaldehyde
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: BJCRWNCKQBPGHU-UHFFFAOYNA-N
SMILES: CC1CC(C)=C(C)CC1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results