Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,6-Trimethyl-3-cyclohexene-1-carboxaldehyde 95.0+%, TCI America™
SDP

Catalog No. T32001ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32001ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32001ML Supplier TCI America Supplier No. T32001ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13702-58-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
InChI Key BJCRWNCKQBPGHU-UHFFFAOYSA-N
PubChem CID 3745161
IUPAC Name 3,4,6-trimethylcyclohex-3-ene-1-carbaldehyde
SMILES CC1CC(=C(CC1C=O)C)C

Specifications

Color Yellow
Boiling Point 93°C
Quantity 1 mL
Formula Weight 152.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4,6-Trimethyl-3-cyclohexene-1-carboxaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.