accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOO

CAS RN 13709-05-2

IUPAC Name: 3-(methoxymethoxy)benzaldehyde
Synonyms: 3-(methoxymethoxy)benzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: JAFJNSQISURLCX-UHFFFAOYSA-N
SMILES: COCOC1=CC=CC(C=O)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results