Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methoxymethoxy)benzaldehyde 95.0+%, TCI America™
SDP

Catalog No. M14485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14485G 5 g
M144825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14485G Supplier TCI America Supplier No. M14485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13709-05-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD04038425
InChI Key JAFJNSQISURLCX-UHFFFAOYSA-N
PubChem CID 12273513
IUPAC Name 3-(methoxymethoxy)benzaldehyde
SMILES COCOC1=CC=CC(=C1)C=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Methoxymethoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.