accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13718-50-8

IUPAC Name: barium(2+) diiodide
Synonyms: barium iodide
Molecular Weight (g/mol): 391.14
Molecular Formula: BaI2
InChi Key: SGUXGJPBTNFBAD-UHFFFAOYSA-L
SMILES: [I-].[I-].[Ba++]
Quantity 
  • (1)
  • (4)
  • (2)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)