Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA4711814
Change view
Click to view available options
Quantity:
25 g
Packaging:
Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4711814 25 g Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4711814 Supplier Thermo Scientific Chemicals Supplier No. 04711814
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13718-50-8 | BaI2 | 391.14 g/mol

Barium iodide is used as a reagent for facile synthesis of poly(phenylcarbyne). It reacts with alkyl potassium compounds to form organobarium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-50-8
Molecular Formula BaI2
Molecular Weight (g/mol) 391.14
MDL Number MFCD00003452
InChI Key SGUXGJPBTNFBAD-UHFFFAOYSA-L
IUPAC Name barium(2+) diiodide
SMILES [I-].[I-].[Ba++]

Specifications

Melting Point 740°C
Color White
Physical Form Beads
Quantity 25 g
UN Number UN1564
Merck Index 14,980
Solubility Information Soluble in water,alcohol,and acetone.
Formula Weight 391.15
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 5.15 g/mL
Assay (metals basis)
Chemical Name or Material Barium iodide, Ultra dry
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 237-276-9
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.