accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1373116-00-7

IUPAC Name: 2,3-difluoro-1-methoxy-4-[(4-propylcyclohexyl)methoxy]benzene
Synonyms: 2,3-difluoro-1-methoxy-4-[(4-propylcyclohexyl)methoxy]benzene
Molecular Weight (g/mol): 298.37
Molecular Formula: C17H24F2O2
InChi Key: VPEMEZFAMQJFFP-UHFFFAOYSA-N
SMILES: CCCC1CCC(COC2=C(F)C(F)=C(OC)C=C2)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results