Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole 98.0+%, TCI America™
SDP

Catalog No. D49095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49095G 5 g
D49091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49095G Supplier TCI America Supplier No. D49095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1373116-00-7
Molecular Formula C17H24F2O2
Molecular Weight (g/mol) 298.37
MDL Number MFCD28976165
InChI Key VPEMEZFAMQJFFP-UHFFFAOYSA-N
Synonym 2,3-Difluoro-1-methoxy-4-[(trans-4-propylcyclohexyl)methoxy]benzene
PubChem CID 57583104
IUPAC Name 2,3-difluoro-1-methoxy-4-[(4-propylcyclohexyl)methoxy]benzene
SMILES CCCC1CCC(COC2=C(F)C(F)=C(OC)C=C2)CC1

Specifications

Melting Point 53°C
Color White
Quantity 5 g
Formula Weight 298.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.