accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3OH3CFFFSOONSOOFFF

CAS RN 1373334-05-4

IUPAC Name: ethyl(3-methoxypropyl)dimethylazanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; ethyl(3-methoxypropyl)dimethylazanium
Molecular Weight (g/mol): 426.39
Molecular Formula: C10H20F6N2O5S2
InChi Key: MBQXUMAOBLRIOQ-UHFFFAOYSA-N
SMILES: CC[N+](C)(C)CCCOC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results