Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl(3-methoxypropyl)dimethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. E12825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12825G Supplier TCI America Supplier No. E12825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1373334-05-4
Molecular Formula C10H20F6N2O5S2
Molecular Weight (g/mol) 426.389
InChI Key MBQXUMAOBLRIOQ-UHFFFAOYSA-N
PubChem CID 132274878
IUPAC Name bis(trifluoromethylsulfonyl)azanide;ethyl-(3-methoxypropyl)-dimethylazanium
SMILES CC[N+](C)(C)CCCOC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 426.39
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl(3-methoxypropyl)dimethylammonium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.