accessibility menu, dialog, popup

UserName

Viewing profile as user:
PP

CAS RN 137349-65-6

IUPAC Name: dicyclohexyl[(dicyclohexylphosphanyl)methyl]phosphane
Synonyms: dicyclohexyl[(dicyclohexylphosphanyl)methyl]phosphane
Molecular Weight (g/mol): 408.59
Molecular Formula: C25H46P2
InChi Key: OWFLJHJHQKHZJR-UHFFFAOYSA-N
SMILES: C(P(C1CCCCC1)C1CCCCC1)P(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results