accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SiH3CCH3NS

CAS RN 137382-38-8

IUPAC Name: 2-(tert-butyldimethylsilyl)-1,3-thiazole
Synonyms: 2-(tert-butyldimethylsilyl)-1,3-thiazole
Molecular Weight (g/mol): 199.39
Molecular Formula: C9H17NSSi
InChi Key: DKQFKRXPJSBPOF-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results